missing translation for 'onlineSavingsMsg'
Learn More
1-[2-(1-Pyrrolidinyl)ethyl]piperazine, 97%, Thermo Scientific™ Product Code.: 11400250

1-[2-(1-Pyrrolidinyl)ethyl]piperazine, 97%, Thermo Scientific™

Product Code. 11400250
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11400250

Brand: Thermo Scientific Alfa Aesar H50692.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-[2-(1-Pyrrolidinyl)ethyl]piperazine is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with air, carbondioxide and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22763-69-5
Molecular Formula C10H21N3
Molecular Weight (g/mol) 183.299
MDL Number MFCD01075190
InChI Key UXHZIFKPLFIPSX-UHFFFAOYSA-N
Synonym 1-2-pyrrolidin-1-yl ethyl piperazine, 1-2-pyrrolidinoethyl piperazine, 1-2-pyrrolidin-1-ylethyl piperazine, 1-2-pyrrolidin-1-yl-ethyl-piperazine, 1-2-1-pyrrolidinyl ethyl piperazine, piperazine, 1-2-1-pyrrolidinyl ethyl, acmc-1cbwl, 2-pyrrolidinylethyl piperazine, 1-2-pyrrolidinoethyl-piperazineShow MoreShow Less
PubChem CID 2737250
IUPAC Name 1-(2-pyrrolidin-1-ylethyl)piperazine
SMILES C1CCN(C1)CCN2CCNCC2

Specifications

Boiling Point 77°C to 79°C (0.5 mmHg)
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 183.3
Percent Purity 97%
Chemical Name or Material 1-[2-(1-Pyrrolidinyl)ethyl]piperazine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberC29S040 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberJ14Q010 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberL20S031 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberC10U047 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberF01S037 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-[2-(1-Pyrrolidinyl)ethyl]piperazine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.