Learn More
1-[2-(Trifluoromethyl)phenyl]pyrrole, 98%, Thermo Scientific™
Product Code. 15467067
Click to view available options
Quantity:
1 g
5 g
25 g
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 89108-30-5
Molecular Formula C11H8F3N
Molecular Weight (g/mol) 211.187
MDL Number MFCD02665223
InChI Key IUMZRHODYBOXLA-UHFFFAOYSA-N
PubChem CID 11367822
IUPAC Name 1-[2-(trifluoromethyl)phenyl]pyrrole
SMILES C1=CC=C(C(=C1)C(F)(F)F)N2C=CC=C2
Specifications
Boiling Point 62°C (1.13 mmHg)
Quantity 5 g
Synonym 1h-pyrrole, 1-2-trifluoromethyl phenyl, 1-2-trifluoromethyl phenyl pyrrole, 1-2-trifluoromethyl phenyl-1h-pyrrole, acmc-20lhrq, 1-2-trifluoromethyl phenyl-1 h-pyrrole
Formula Weight 211.19
Percent Purity 98%
Chemical Name or Material 1-[2-(Trifluoromethyl)phenyl]pyrrole