missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 1,3,4,6-Tetra-O-acetyl-beta-D-glucosamine hydrochloride
Click to view available options
Quantity:
1 g
5 g
25 g
50 g
Description
NSC 82044 is a potential metabolic inhibitor of cellular-membrane glycoconjugates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
NSC 82044 is a potential metabolic inhibitor of cellular-membrane glycoconjugates.
Solubility
Soluble in water at 10mg/ml
Notes
Keep container tightly closed in a dry and well-ventilated place. Store in cool place.
NSC 82044 is a potential metabolic inhibitor of cellular-membrane glycoconjugates.
Solubility
Soluble in water at 10mg/ml
Notes
Keep container tightly closed in a dry and well-ventilated place. Store in cool place.

Specifications
Specifications
Color | Brown to White |
Quantity | 25 g |
Solubility Information | Soluble in water at 10mg/ml |
Formula Weight | 383.78 |
Physical Form | Powder |
Chemical Name or Material | 1,3,4,6-Tetra-O-acetyl-beta-D-glucosamine hydrochloride |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals 1,3,4,6-Tetra-O-acetyl-beta-D-glucosamine hydrochloride >
Spot an opportunity for improvement?Share a Content Correction