missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Allyl-4-hydroxyphenyl)ethan-1-one, 97%, Thermo Scientific™

Product Code. 10344532
10 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1132-05-4
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00068015
InChI Key XVTCWUFLNLZPEJ-UHFFFAOYSA-N
PubChem CID 259653
IUPAC Name 1-(4-hydroxy-3-prop-2-enylphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)O)CC=C

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym 3'-allyl-4'-hydroxyacetophenone, acetophenone, 3'-allyl-4'-hydroxy, 1-3-allyl-4-hydroxyphenyl ethan-1-one, 1-3-allyl-4-hydroxyphenyl ethanone, ethanone, 1-4-hydroxy-3-2-propenyl phenyl, 1-4-hydroxy-3-prop-2-en-1-yl phenyl ethanone, ethanone,1-4-hydroxy-3-2-propen-1-yl phenyl, ethanone, 1-4-hydroxy-3-2-propen-1-yl phenyl, zlchem 1234
Formula Weight 176.21
Percent Purity 95%
Chemical Name or Material 1-(3-Allyl-4-hydroxyphenyl)ethan-1-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(3-Allyl-4-hydroxyphenyl)ethan-1-one, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.