missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxan-6-carboxaldehyde, 98%, Thermo Scientific™

Product Code. 10103404
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 29668-44-8
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00010092
InChI Key CWKXDPPQCVWXAG-UHFFFAOYSA-N
Synonym 1,4-benzodioxan-6-carboxaldehyde, 1,4-benzodioxan-6-carbaldehyde, 2,3-dihydrobenzo b 1,4 dioxine-6-carbaldehyde, 1,4-benzodioxane-6-carboxaldehyde, 3,4-ethylenedioxybenzaldehyde, 1,4-benzodioxin-6-carboxaldehyde, 2,3-dihydro, 7-formyl-1,4-benzodioxane, 1,4-benzodioxane-6-aldehyde, 2,3-dihydro-1,4-benzodioxin-6-carbaldehyde, 1,4-benzodioxane-6-carbaldehyde
PubChem CID 248127
IUPAC Name 2,3-dihydro-1,4-benzodioxine-6-carbaldehyde
SMILES C1COC2=C(O1)C=CC(=C2)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 52.0°C to 53.0°C
Color Beige-Yellow
Boiling Point 105°C (20.0 hPa)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 164.16
Percent Purity 98%
Physical Form Chunks and/or Powder
Chemical Name or Material 1, 4-Benzodioxan-6-carboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Benzodioxan-6-carboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'