missing translation for 'onlineSavingsMsg'
Learn More
1-(4-Bromophenyl)-3-dimethylamino-2-propen-1-one, 95%, Thermo Scientific™ Product Code.: 15498825

1-(4-Bromophenyl)-3-dimethylamino-2-propen-1-one, 95%, Thermo Scientific™

Product Code. 15498825
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 15498825

Brand: Thermo Scientific Alfa Aesar H34204.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73387-60-7
Molecular Formula C11H12BrNO
Molecular Weight (g/mol) 254.13
MDL Number MFCD00121187
InChI Key IGZLESKZUATMSD-UHFFFAOYSA-N
Synonym 2e-1-4-bromophenyl-3-dimethylamino prop-2-en-1-one, 1-4-bromophenyl-3-dimethylaminopropenone, e-1-4-bromophenyl-3-dimethylamino prop-2-en-1-one, 1-4-bromophenyl-3-dimethylamino prop-2-en-1-one, 1-4-bromophenyl-3-dimethylamino-2-propen-1-one, 4-bromophenyl 2-dimethylamino ethenyl ketone, e-1-4-bromophenyl-3-dimethylaminopropenone, e-4'-bromo-beta-dimethylamino acrylophenone, 2e-1 4-bromophenyl-3-dimethylamino prop-2-en-1-one, 2e-1-4-bromophenyl-3-dimethylamino-2-propen-1-one
PubChem CID 5376915
IUPAC Name (E)-1-(4-bromophenyl)-3-(dimethylamino)prop-2-en-1-one
SMILES CN(C)C=CC(=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 80°C to 82°C
Quantity 10 g
Formula Weight 254.13
Percent Purity 95%
Chemical Name or Material 1-(4-Bromophenyl)-3-dimethylamino-2-propen-1-one

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10167086 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(4-Bromophenyl)-3-dimethylamino-2-propen-1-one, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.