missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diethylbenzene, 98%

Product Code. 11400973
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11400973

Brand: Thermo Scientific Alfa Aesar A13343.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 105-05-5 | C10H14 | 134.22 g/mol

1,4-Diethylbenzene, is used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics andcross-linking agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diethylbenzene, is used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics andcross-linking agents.

Solubility
It is insoluble in water. But soluble in ethanol, benzene,carbon tetrachloride.

Notes
Stable under ordinary conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-05-5
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00009264
InChI Key DSNHSQKRULAAEI-UHFFFAOYSA-N
Synonym p-diethylbenzene, benzene, 1,4-diethyl, p-ethylethylbenzene, benzene, p-diethyl, para-diethylbenzene, unii-0psm16x42d, acmc-1bu3z, dsstox_cid_6711, dsstox_rid_78192, dsstox_gsid_26711
PubChem CID 7734
ChEBI CHEBI:34062
IUPAC Name 1,4-diethylbenzene
SMILES CCC1=CC=C(CC)C=C1

Specifications

Melting Point -43°C
Density 0.862
Boiling Point 182°C to 184°C
Flash Point 55°C (131°F)
Odor Aromatic
Refractive Index 1.495
Quantity 100 g
UN Number UN2049
Beilstein 1903396
Solubility Information It is insoluble in water. But soluble in ethanol,benzene,carbon tetrachloride.
Formula Weight 134.22
Percent Purity 98%
Chemical Name or Material 1,4-Diethylbenzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.