missing translation for 'onlineSavingsMsg'
Learn More

1-(5-(Trifluoromethyl)-2-pyridyl)piperidine-4-carboxylic acid, 95%, Thermo Scientific™

40.30 € - 573.00 €

Chemical Identifiers

CAS 406476-31-1
Molecular Formula C12H13F3N2O2
Molecular Weight (g/mol) 274.24
MDL Number MFCD00209248
InChI Key KNDSIDUPVUCATQ-UHFFFAOYSA-N
Synonym 1-5-trifluoromethyl pyridin-2-yl piperidine-4-carboxylic acid, 1-5-trifluoromethyl-2-pyridyl piperidine-4-carboxylic acid, 5'-trifluoromethyl-3,4,5,6-tetrahydro-2h-1,2' bipyridinyl-4-carboxylic acid, peakdale1_002677, 2-4-carboxypiperidin-1-yl-5-trifluoromethyl pyridine, 4-carboxy-1-5-trifluoromethyl pyridin-2-yl piperidine, 1-5-trifluoromethyl pyridin-2-yl piperidine-4-carboxylicacid, 1-5-trifluoromethylpyrid-2-yl-piperidine-4-carboxylic acid
PubChem CID 2777788
SMILES [O-]C(=O)C1CCN(CC1)C1=CC=C(C=[NH+]1)C(F)(F)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10350383
SDS View Documents
Thermo Scientific Maybridge
KM07694CB
250 mg
40.30 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10106792
SDS View Documents
Thermo Scientific Maybridge
KM07694DA
1 g
108.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10317164
SDS View Documents
Thermo Scientific Maybridge
KM07694EA
10 g
573.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(5-(Trifluoromethyl)-2-pyridyl)piperidine-4-carboxylic acid, 95%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.