Learn More
1-BOC-3-(aminomethyl)azetidine, 95%, Thermo Scientific™
94.93 € - 277.00 €
Chemical Identifiers
CAS | 325775-44-8 |
---|---|
Molecular Formula | C9H18N2O2 |
Molecular Weight (g/mol) | 186.25 |
InChI Key | XSJPKMUFBHSIRA-UHFFFAOYSA-N |
Synonym | 1-boc-3-aminomethyl azetidine, tert-butyl 3-aminomethyl azetidine-1-carboxylate, 1-boc-3-aminomethylazetidine, 3-aminomethyl-1-n-boc-azetidine, 1-n-boc-3-aminomethylazetidine, 3-aminomethyl-azetidine-1-carboxylic acid tert-butyl ester, 1-boc-3-aminomethyl-azetidine, 3-aminomethyl azetidine, n1-boc protected, tert-butyl 3-aminomethyl azetidinecarboxylate, 1-azetidinecarboxylic acid, 3-aminomethyl-, 1,1-dimethylethyl esterShow More |
PubChem CID | 10679125 |
IUPAC Name | tert-butyl 3-(aminomethyl)azetidine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CC(C1)CN |
Description
Chemical Identifiers
325775-44-8 | |
186.25 | |
1-boc-3-aminomethyl azetidine, tert-butyl 3-aminomethyl azetidine-1-carboxylate, 1-boc-3-aminomethylazetidine, 3-aminomethyl-1-n-boc-azetidine, 1-n-boc-3-aminomethylazetidine, 3-aminomethyl-azetidine-1-carboxylic acid tert-butyl ester, 1-boc-3-aminomethyl-azetidine, 3-aminomethyl azetidine, n1-boc protected, tert-butyl 3-aminomethyl azetidinecarboxylate, 1-azetidinecarboxylic acid, 3-aminomethyl-, 1,1-dimethylethyl esterShow More | |
tert-butyl 3-(aminomethyl)azetidine-1-carboxylate |
C9H18N2O2 | |
XSJPKMUFBHSIRA-UHFFFAOYSA-N | |
10679125 | |
CC(C)(C)OC(=O)N1CC(C1)CN |
Specifications
325775-44-8 | |
C9H18N2O2 | |
XSJPKMUFBHSIRA-UHFFFAOYSA-N | |
tert-butyl 3-(aminomethyl)azetidine-1-carboxylate | |
>110°C | |
186.25 | |
1-BOC-3-(aminomethyl)azetidine |
94% min. (GC) | |
1-boc-3-aminomethyl azetidine, tert-butyl 3-aminomethyl azetidine-1-carboxylate, 1-boc-3-aminomethylazetidine, 3-aminomethyl-1-n-boc-azetidine, 1-n-boc-3-aminomethylazetidine, 3-aminomethyl-azetidine-1-carboxylic acid tert-butyl ester, 1-boc-3-aminomethyl-azetidine, 3-aminomethyl azetidine, n1-boc protected, tert-butyl 3-aminomethyl azetidinecarboxylate, 1-azetidinecarboxylic acid, 3-aminomethyl-, 1,1-dimethylethyl ester | |
CC(C)(C)OC(=O)N1CC(C1)CN | |
186.25 | |
10679125 | |
95% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air
GHS Signal Word: Warning
- 1-Boc-3-(aminomethyl)azetidine, 95%
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.