Learn More
1-Hexadecanethiol, 97% (dry wt.), may cont. up to 4% water
Description
1-Hexadecanethiol is used in the study of nanoparticles and self-assembled monolayers. It is also used to prepare thiol-functionalized 1.5 nm gold nanoparticles via a ligand exchange process with triphenylphosphine-stabilized nanoparticles and in the formation of hydrophobic self-assembled monolayers (SAMs). Due to the length of the alkane chain, the resulting monolayer is highly ordered. 1-Hexadecanethiol is also employed in different fields such as biomedicine, chemical industry, also used in electroplating industry and chemical auxiliaries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Hexadecanethiol is used in the study of nanoparticles and self-assembled monolayers. It is also used to prepare thiol-functionalized 1.5 nm gold nanoparticles via a ligand exchange process with triphenylphosphine-stabilized nanoparticles and in the formation of hydrophobic self-assembled monolayers (SAMs). Due to the length of the alkane chain, the resulting monolayer is highly ordered. 1-Hexadecanethiol is also employed in different fields such as biomedicine, chemical industry, also used in electroplating industry and chemical auxiliaries.
Solubility
Soluble in methanol, benzene, ether, acetone and ethyl acetate. Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under inert gas. It is sensitive to air. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 19°C to 23°C |
| Density | 0.846 |
| Boiling Point | 334°C |
| Flash Point | 135°C (275°F) |
| Odor | Unpleasant |
| Linear Formula | CH3(CH2)15SH |
| Refractive Index | 1.4625 |
| Quantity | 100 mL |
| Beilstein | 1748495 |
| Sensitivity | Air Sensitive |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.