Learn More
1-Phenyl-1-pentyne, 98%, Thermo Scientific Chemicals
31.76 € - 101.51 €
Chemical Identifiers
CAS | 4250-81-1 |
---|---|
Molecular Formula | C11H12 |
Molecular Weight (g/mol) | 144.22 |
MDL Number | MFCD00039959 |
InChI Key | DEGIOKWPYFOHGH-UHFFFAOYSA-N |
Synonym | 1-phenyl-1-pentyne, pent-1-yn-1-ylbenzene, 1-pentynylbenzene, benzene, 1-pentynyl, 1-pentynylbenzene #, acmc-1aso7, 1-phenyl-2-propylacetylene |
PubChem CID | 77923 |
IUPAC Name | pent-1-ynylbenzene |
SMILES | CCCC#CC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11485806
|
Thermo Scientific Alfa Aesar
B20857.06 |
5 g |
31.76 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11495806
|
Thermo Scientific Alfa Aesar
B20857.14 |
25 g |
101.51 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It finds it application in the highly regioselective synthesis of polysubstituted naphthalene derivatives through gallium trichloride catalyzed alkyne-aldehyde coupling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds it application in the highly regioselective synthesis of polysubstituted naphthalene derivatives through gallium trichloride catalyzed alkyne-aldehyde coupling.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
4250-81-1 | |
144.22 | |
DEGIOKWPYFOHGH-UHFFFAOYSA-N | |
77923 | |
CCCC#CC1=CC=CC=C1 |
C11H12 | |
MFCD00039959 | |
1-phenyl-1-pentyne, pent-1-yn-1-ylbenzene, 1-pentynylbenzene, benzene, 1-pentynyl, 1-pentynylbenzene #, acmc-1aso7, 1-phenyl-2-propylacetylene | |
pent-1-ynylbenzene |
Specifications
4250-81-1 | |
213°C to 215°C | |
C11H12 | |
1.5413 | |
1853156 | |
Not miscible or difficult to mix in water. | |
CCCC#CC1=CC=CC=C1 | |
144.22 | |
144.22 | |
1-Phenyl-1-pentyne |
0.906 | |
82°C (179°F) | |
CH3CH2CH2C≡CC6H5 | |
MFCD00039959 | |
1-phenyl-1-pentyne, pent-1-yn-1-ylbenzene, 1-pentynylbenzene, benzene, 1-pentynyl, 1-pentynylbenzene #, acmc-1aso7, 1-phenyl-2-propylacetylene | |
DEGIOKWPYFOHGH-UHFFFAOYSA-N | |
pent-1-ynylbenzene | |
77923 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'einecsNumber' : 224-211-4
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.