Learn More
uvex 1 Sport White NC Shoe O1 FO SRC, Width 11
With a lightweight design, high-tech materials, and ergonomically designed outsole, the Uvex 1 Sport White NC Shoe is the ideal companion for the working day and beyond. The hydrophobic upper ensures optimized climate control, and reliably protects the wearer against liquids and other contaminants.
130.00 €
Specifications
Color | White |
---|---|
Material | Textile |
Toe Material | Plain Toe |
Certifications/Compliance (Europe) | EN ISO 20347:2012 O1 FO SRC EN ISO 20347:2022 O1 FO SC SR |
Gender | Unisex |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17843170
|
uvex
6570835 |
35 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17853170
|
uvex
6570836 |
36 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17863170
|
uvex
6570837 |
37 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17873170
|
uvex
6570838 |
38 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17883170
|
uvex
6570839 |
39 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17893170
|
uvex
6570840 |
40 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17803180
|
uvex
6570841 |
41 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17813180
|
uvex
6570842 |
42 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17823180
|
uvex
6570843 |
43 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17833180
|
uvex
6570844 |
44 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17843180
|
uvex
6570845 |
45 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17853180
|
uvex
6570846 |
46 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17863180
|
uvex
6570847 |
47 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17873180
|
uvex
6570848 |
48 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17883180
|
uvex
6570849 |
49 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17893180
|
uvex
6570850 |
50 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17803190
|
uvex
6570851 |
51 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17813190
|
uvex
6570852 |
52 |
130.00 €
1 pair |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
Description
- Modern, ultra-lightweight and flexible O1 occupational shoe without protective toe cap
- Washable at up to 30°C
- Made from synthetic materials, so suitable for people allergic to chrome
- Elastic laces with quick release for individual adjustment
- All sole materials are free of silicone, plasticizers and other substances that interfere with wetting agents
- New, innovative uvex i-PUREnrj sole technology provides outstanding shock-absorption properties at the forefoot and heel and excellent rebound energy over the entire midsole
- Meets the ESD specification with volume resistance less than 35 Megohms
- Ergonomically designed outsole made from dual density polyurethane with excellent slip resistance
- Outstanding wearer comfort thanks to a newly developed last and climate-optimized, breathable materials
- Virtually seam-free construction with high-tech materials to eliminate pressure points
- Removable anti-static comfortable insole with a moisture transport system and additional shock absorption in the forefoot and heel
- Soft padding on collar and tongue
- Made partly from recycled materials in order to conserve resources
- Free from any of the harmful substances listed on our banned hazardous substances list
Specifications
White | |
Plain Toe | |
Unisex | |
Yes | |
No |
Textile | |
EN ISO 20347:2012 O1 FO SRC EN ISO 20347:2022 O1 FO SC SR | |
Standard Width | |
Trainers | |
ESD Safety Shoe |