Learn More
Masterflex™ Stopcocks with Luer Connections
Regulate flow without any leaking using these fittings. Cole-Parmer™ Stopcocks with Luer Connections are available with either a male luer lock or a male luer slip.
Brand: Masterflex™ 30600-01
Description
Regulate flow without any leaking using these fittings. Cole-Parmer™ Stopcocks with Luer Connections are available with either a male luer lock or a male luer slip.
- Handle rotates 90°
- PC body and HDPE stem
Specifications
Stopcock | |
Polycarbonate, HDPE | |
Polycarbonate and HDPE | |
Male Luer Slip |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Promotions
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Masterflex™ Stopcocks with Luer Connections > Luer, Male slip
Spot an opportunity for improvement?Share a Content Correction