missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Refractory Elements Standard: Al, B, Cr, Hf, Mo, Nb, Si, Ta, Ti, V, W, Zr @ 100 μg/mL in 5% HCl, tr. HF, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
Refractory Elements Standard: Al, B, Cr, Hf, Mo, Nb, Si, Ta, Ti, V, W, Zr @ 100 μg/mL in 5% HCl, tr. HF
Specifications
Specifications
| Chemical Name or Material | Aluminium/Silicon/Boron/Molybdenum/Chromium/Hafnium/Niobium/Tantalum/Titanium/Vanadium/Zirconium/Tungsten |
| CAS | 7429-90-5, 7440-21-3, 7440-42-8, 7439-98-7, 7440-47-3, 7440-58-6 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Aluminium/Silicon/Boron/Molybdenum/Chromium/Hafnium/Niobium/Tantalum/Titanium/Vanadium/Zirconium/Tungsten |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HCl |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Aluminium 99.95 μg/mL/Silicon 99.98 μg/mL/Boron 99.99 μg/mL/Molybdenum 99.99 μg/mL/Chromium 100 μg/mL/Hafnium 100 μg/mL/Niobium 100 μg/mL/Tantalum 100 μg/mL/Titanium 100 μg/mL/Vanadium 100 μg/mL/Zirconium 100 μg/mL/Tungsten 100.1 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction