Learn More
Fisher Chemical Sodium Chloride, Certified AR for Analysis, 10kg
Brand:  Fisher Chemical S/3160/68
Sodium Chloride, Certified AR for Analysis, 10kg
Product Code. 10194763
  • ONLINE EXCLUSIVE PRICE
    167.00 €
    164.00 € / 10kg
    Save 3.00 €
    1% Off