Learn More
Bochem™ 18/10 Stainless Steel Double Ended Micro-spatulas
Product Code. 10507892
Click to view available options
Length (Metric):
100 mm
130 mm
150 mm
185 mm
210 mm
Length (Metric) Blade:
30 mm
35 mm
40 mm
50 mm
55 mm
Width (Metric) Blade:
2 mm
3 mm
4 mm
5 mm
6 mm
9 mm
Unit Size:
Each
Product Code. 10507892
Brand: Bochem™ 3015
Only null left
Add to basket
Add to basket
Manufactured from stainless steel. Bochem™ 18/10 Stainless Steel Double Ended Micro-spatulas are multi-purpose.
  • Double endet
  • Stainless steel
  • Autoclavable
  • Straight
Specifications
Quantity 1 Ea.
Autoclavable Autoclavable
Shape Straight
Product Type Double Ended Micro Spatula
For Use With (Application) Multi-purpose spatula for a wide range of uses
Length (Metric) Blade 40 mm
Width (Metric) 3 mm
Width (Metric) Blade 3 mm
Blade Material Stainless Steel (18/10)
Diameter (Metric) Outer 1.5mm
Dimensions (L x W) 150 x 3 mm
Material Stainless Steel (18/10)
Length (Metric) 150 mm