Learn More
Bochem™ 18/10 Stainless Steel Precision Forceps
Click to view available options
Length (Metric):
105 mm
130 mm
Description:
Curved
Straight
Unit Size:
Each
Description
- Precision forceps
- Extra sharp
- Points without ridges
Specifications
Specifications
| Tip Style | Extra sharp |
| Material | Stainless Steel (18/10) |
| Length (Metric) | 105 mm |
| Product Type | Forceps |
| Quantity (No. per Case) | Each |
| Precision | Yes |
| Type | Forceps, Precision |
| Description | Curved |