Learn More
1A-116, MedChemExpress
MedChemExpress 1A-116, a potent Rac1 inhibitor, is specific for W56 residues, can prevent EGF-induced Rac1 activation and block Rac1-P-Rex1 interaction. 1A-116 can induce apoptosis and inhibit cell proliferation, migration and cycle progression in a concentration-dependent manner. 1A-116 also demonstrates a high antimetastatic activity in vivo.

195.75 € - 1185.30 €
Specifications
Chemical Name or Material | 1A-116 |
---|---|
Physical Form | Solid |
CAS | 1430208-73-3 |
Color | Off-White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16474966
![]() |
Medchem Express
HY-104064 |
100 mg |
1185.30 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16453948
![]() |
Medchem Express
HY-104064 |
10 mg |
291.60 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16442926
![]() |
Medchem Express
HY-104064 |
25 mg |
525.15 €
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16436667
![]() |
Medchem Express
HY-104064 |
5 mg |
195.75 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16410307
![]() |
Medchem Express
HY-104064 |
50 mg |
789.75 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1A-116, a potent Rac1 inhibitor, is specific for W56 residues, can prevent EGF-induced Rac1 activation and block Rac1-P-Rex1 interaction. 1A-116 can induce apoptosis and inhibit cell proliferation, migration and cycle progression in a concentration-dependent manner. 1A-116 also demonstrates a high antimetastatic activity in vivo.Specifications
1A-116 | |
1430208-73-3 | |
Research | |
Cancer-Kinase/protease | |
N=C(NC1=CC=CC=C1C(F)(F)F)NC2=CC(C)=CC(C)=C2 | |
307.31 | |
Research |
Solid | |
Off-White | |
C16H16F3N3 | |
DMSO : 100 mg/mL (325.40 mM; Need ultrasonic) | |
307.31 | |
99.39% |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.