missing translation for 'onlineSavingsMsg'
Learn More
1H,1H-Perfluorooctylamine, 97%, Thermo Scientific™ Product Code.: 11373786

1H,1H-Perfluorooctylamine, 97%, Thermo Scientific™

Product Code. 11373786
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11373786

Brand: Thermo Scientific Alfa Aesar L16845.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1H,1H-Perfluorooctylamine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307-29-9
Molecular Formula C8H4F15N
Molecular Weight (g/mol) 399.103
MDL Number MFCD00042460
InChI Key HZCZXRSVKNFILZ-UHFFFAOYSA-N
PubChem CID 136148
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctan-1-amine
SMILES C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)N

Specifications

Density 1.714
Boiling Point 149°C to 150°C
Flash Point None
Odor Amine-like
Linear Formula CF3(CF2)6CH2NH2
Refractive Index 1.305
Quantity 1 g
UN Number UN2735
Beilstein 1806966
Synonym 1h,1h-perfluorooctylamine, 1h,1h-pentadecafluorooctylamine, 1-octanamine, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro, pentadecafluorooctylamine, 1h,1h-pentadecafluorooctylamin, 1h,1h-perfluorooctylamine 1g, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-octylamine, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctylamine, 1-octanamine,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 399.1
Percent Purity 97%
Chemical Name or Material 1H,1H-Perfluorooctylamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10197935 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot Number10206901 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberUSLF004962 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberLF004962 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot Number10180343 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1H,1H-Perfluorooctylamine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.