missing translation for 'onlineSavingsMsg'
Learn More
(1S,3R)-cis-3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid, 97%, Thermo Scientific Chemicals Product Code.: 15400096

(1S,3R)-cis-3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15400096
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15400096

Brand: Thermo Scientific Alfa Aesar H52747.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96443-42-4
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD04972280
InChI Key FVUHGTQDOMGZOT-NTSWFWBYSA-N
Synonym 1s,3r-3-methoxycarbonyl cyclopentanecarboxylic acid, 1s,3r-3-methoxycarbonyl cyclopentane-1-carboxylic acid, 1r,3s-1,3-cyclopentanedicarboxylic acid 1-methyl ester, 1s,3r-cis-3-carbomethoxy cyclopentane-1-carboxylic acid, cis-3-methoxycarbonyl cyclopentancarboxylic acid, 1s,3r-cis-3-carbomethoxy cyclopentane-1-carboxyl, 1,3-cyclopentanedicarboxylicacid,1-methyl ester, 1r,3s, 1r-cis-1,3-cyclopentanedicarboxylic acid monomethyl ester, 1,3-cyclopentanedicarboxylic acid, 1-methyl ester, 1r,3s
PubChem CID 10888375
IUPAC Name (1S,3R)-3-methoxycarbonylcyclopentane-1-carboxylic acid
SMILES COC(=O)C1CCC(C1)C(=O)O

Specifications

Quantity 1 g
Formula Weight 172.18
Percent Purity 97%
Chemical Name or Material (1S,3R)-cis-3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY16B019 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52747.MD,H52747.03
Lot NumberI03Z057 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52747.MD,H52747.03
Lot NumberQ29A018 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52747.MD,H52747.03
Lot NumberK02W055 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52747.MD,H52747.03
Lot NumberC14Z023 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52747.MD,H52747.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(1S,3R)-cis-3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.