missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,4R)-2-Azabicyclo[2.2.1]heptan-3-one, 95%, 98% ee, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
Specifications
Specifications
| Melting Point | 76.2°C |
| Color | White to Yellow |
| Enantiomeric Excess | 97.5% min. |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94% min. (GC) |
| Packaging | Glass bottle |
| Molecular Formula | C6H9NO |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 111.14 |
| Formula Weight | 111.14 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction