missing translation for 'onlineSavingsMsg'
Learn More

(1S,4R)-2-Azabicyclo[2.2.1]heptan-3-one, 95%, 98% ee, Thermo Scientific™

Product Code. 11966621 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11966621

Brand: Thermo Scientific Acros 362220050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 134003-03-5
InChI Key UIVLZOWDXYXITH-UHNVWZDZSA-N
Synonym 1s,4r-2-azabicyclo 2.2.1 heptan-3-one, 2-azabicyclo 2.2.1 heptan-3-one, 1s,4r, 1s,4r-2-aza-bicyclo 2.2.1 heptan-3-one, 1beta,4beta-2-azabicyclo 2.2.1 heptane-3-one
PubChem CID 2734523
IUPAC Name (1R,4S)-3-azabicyclo[2.2.1]heptan-2-one
SMILES C1CC2CC1C(=O)N2

Specifications

Color White to Yellow
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C6H9NO
Quantity 5 g
Molecular Weight (g/mol) 111.144
Formula Weight 111.14
Percent Purity ≥94% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1S, 4R)-2-Azabicyclo[2.2.1]heptan-3-one
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.