missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Adamantyl)-4-methylphenol, 99%, Thermo Scientific Chemicals

30.60 € - 304.52 €

Chemical Identifiers

CAS 41031-50-9
Molecular Formula C17H22O
Molecular Weight (g/mol) 242.36
MDL Number MFCD00168147
InChI Key XHLJIHBDAJFXBE-UHFFFAOYSA-N
Synonym 2-1-adamantyl-4-methylphenol, 2-adamantan-1-yl-4-methylphenol, pubchem21690, acmc-1anoj, 2-1-adamantyl-4-methyl-phenol, 2-adamantan-1-yl-4-methyl-phenol, phenol,4-methyl-2-tricyclo 3.3.1.13,7 dec-1-yl, 4-methyl-2-3r,5s,7s-adamantan-1-yl phenol, phenol, 4-methyl-2-tricyclo 3.3.1.1 3,7 dec-1-ylShow MoreShow Less
PubChem CID 617992
IUPAC Name 2-(1-adamantyl)-4-methylphenol
SMILES CC1=CC(=C(O)C=C1)C12CC3CC(CC(C3)C1)C2
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15472318
SDS View Documents
Thermo Scientific Alfa Aesar
H55922.MD
250 mg N/A N/A N/A
15452318
SDS View Documents
Thermo Scientific Alfa Aesar
H55922.03
1 g
86.03 €
1g
Please to purchase this item. Need a web account? Register with us today!
15462318
SDS View Documents
Thermo Scientific Alfa Aesar
H55922.06
5 g
304.52 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-(1-Adamantyl)-4-methylphenol, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(1-Adamantyl)-4-methylphenol, is used as a pharmaceutical intermediate.

Solubility
Sparingly Soluble in water (7.8E-3 g/L) (25°C).

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(1-Adamantyl)-4-methylphenol, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.