missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Naphthyl)-5-phenyloxazole, laser grade and suitable for scintillation spectrometry, 99+%, Thermo Scientific Chemicals

37.29 € - 106.53 €

Chemical Identifiers

CAS 846-63-9
Molecular Formula C19H13NO
Molecular Weight (g/mol) 271.319
MDL Number MFCD00019714
InChI Key WWVFJJKBBZXWFV-UHFFFAOYSA-N
Synonym 2-1-naphthyl-5-phenyloxazole, alpha-npo, oxazole, 2-1-naphthalenyl-5-phenyl, 2-1-naphthyl-5-phenyl-1,3-oxazole, 2-naphthalen-1-yl-5-phenyloxazole, oxazole, 2-1-naphthyl-5-phenyl, .alpha.-npo, anpo, 2-alpha-naphthyl-5-phenyloxazole, 2-naphthalen-1-yl-5-phenyl-1,3-oxazole
PubChem CID 70058
IUPAC Name 2-naphthalen-1-yl-5-phenyl-1,3-oxazole
SMILES C1=CC=C(C=C1)C2=CN=C(O2)C3=CC=CC4=CC=CC=C43
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11409230
SDS View Documents
Thermo Scientific Alfa Aesar
A16679.03
1 g
37.29 €
1g
Please to purchase this item. Need a web account? Register with us today!
11419230
SDS View Documents
Thermo Scientific Alfa Aesar
A16679.06
5 g
106.53 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Starting with the organic fluor 2-(1-naphthyl)-5-phenyloxazole (αNPO), the vNPO and allylNPO monomers were synthesized using Stille coupling followed by purification. Used as a laser dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Starting with the organic fluor 2-(1-naphthyl)-5-phenyloxazole (αNPO), the vNPO and allylNPO monomers were synthesized using Stille coupling followed by purification. Used as a laser dye.

Solubility
Sparingly Soluble (4.1E-4 g/L) (25°C). Soluble most organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 212-689-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(1-Naphthyl)-5-phenyloxazole, laser grade and suitable for scintillation spectrometry, 99+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.