missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, ≥99.0% (GC), Honeywell™

Product Code. 15645880
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Purum,≥99.0% (GC)

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

CAS 540-84-1
Density 0.692g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.06
Flash Point −12°C (10.4°F)
Ignition Point 745°F
Packaging Glass Bottle
Molecular Formula C8H18
Linear Formula (CH3)2CHCH2C(CH3)3
MDL Number MFCD00008943
Vapor Pressure 41mmHg (21°C)
UN Number UN1262
Beilstein 1696876
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILES CC(C)CC(C)(C)C
IUPAC Name 2,2,4-trimethylpentane
Molecular Weight (g/mol) 114.232
PubChem CID 10907
ChEBI CHEBI:62805
Formula Weight 114.23g/mol
Percent Purity ≥99.0% (GC)
Chemical Name or Material 2,2,4-Trimethylpentane
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'