Learn More
2,2-Dimethyl-4-pentenoic acid, 95%, Thermo Scientific Chemicals
71.56 € - 248.24 €
Chemical Identifiers
CAS | 16386-93-9 |
---|---|
Molecular Formula | C7H12O2 |
Molecular Weight (g/mol) | 128.17 |
MDL Number | MFCD00012304 |
InChI Key | BGUAPYRHJPWVEM-UHFFFAOYSA-N |
Synonym | 2,2-dimethyl-4-pentenoic acid, 4-pentenoic acid, 2,2-dimethyl, 2,2-dimethyl-pent-4-enoic acid, 2,2-dimethyl-4-pentenoicacid, pubchem12797, acmc-1c5lb, ksc495e3r, 2,2-dimethyl4-pentenoic acid, 2,2-dimethyl-1-4-pentenoic acid 1g |
PubChem CID | 140065 |
IUPAC Name | 2,2-dimethylpent-4-enoic acid |
SMILES | CC(C)(CC=C)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15477438
|
Thermo Scientific Alfa Aesar
H60940.03 |
1 g |
71.56 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15487438
|
Thermo Scientific Alfa Aesar
H60940.06 |
5 g |
248.24 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents. Store under room temperature.
Chemical Identifiers
16386-93-9 | |
128.17 | |
BGUAPYRHJPWVEM-UHFFFAOYSA-N | |
140065 | |
CC(C)(CC=C)C(O)=O |
C7H12O2 | |
MFCD00012304 | |
2,2-dimethyl-4-pentenoic acid, 4-pentenoic acid, 2,2-dimethyl, 2,2-dimethyl-pent-4-enoic acid, 2,2-dimethyl-4-pentenoicacid, pubchem12797, acmc-1c5lb, ksc495e3r, 2,2-dimethyl4-pentenoic acid, 2,2-dimethyl-1-4-pentenoic acid 1g | |
2,2-dimethylpent-4-enoic acid |
Specifications
16386-93-9 | |
104°C to 108°C | |
C7H12O2 | |
1.431 | |
UN3265 | |
Soluble in water. | |
CC(C)(CC=C)C(O)=O | |
128.17 | |
128.17 | |
2,2-Dimethyl-4-pentenoic acid |
0.933 | |
88°C (190°F) | |
H2C=CHCH2C(CH3)2CO2H | |
MFCD00012304 | |
2,2-dimethyl-4-pentenoic acid, 4-pentenoic acid, 2,2-dimethyl, 2,2-dimethyl-pent-4-enoic acid, 2,2-dimethyl-4-pentenoicacid, pubchem12797, acmc-1c5lb, ksc495e3r, 2,2-dimethyl4-pentenoic acid, 2,2-dimethyl-1-4-pentenoic acid 1g | |
BGUAPYRHJPWVEM-UHFFFAOYSA-N | |
2,2-dimethylpent-4-enoic acid | |
140065 | |
95% |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.