missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Tri-O-benzyl-beta-D-arabinopyranose, 97+%, Thermo Scientific™

Product Code. 11329885
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11329885

Brand: Thermo Scientific Alfa Aesar L19870.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18039-26-4
Molecular Formula C26H28O5
Molecular Weight (g/mol) 420.505
MDL Number MFCD05664045
InChI Key HTSKDJMXBBFKKG-FXSWLTOZSA-N
Synonym 2,3,4-tri-o-benzyl-beta-d-arabinopyranose, 2r,3s,4r,5r-3,4,5-tris benzyloxy tetrahydro-2h-pyran-2-ol, 2r,3s,4r,5r-3,4,5-tris benzyloxy oxan-2-ol, 2r,3s,4r,5r-3,4,5-tris phenylmethoxy oxan-2-ol, 2,3,4-tri-o-benzyl-b-d-arabinopyranose, arabinopyranose,2,3,4-tri-o-benzyl-,b-d-8ci
PubChem CID 7578140
IUPAC Name (2R,3S,4R,5R)-3,4,5-tris(phenylmethoxy)oxan-2-ol
SMILES C1C(C(C(C(O1)O)OCC2=CC=CC=C2)OCC3=CC=CC=C3)OCC4=CC=CC=C4

Specifications

Melting Point 73°C to 75°C
Quantity 250 mg
Beilstein 5303314
Optical Rotation −70° (c=1 in Methanol)
Formula Weight 420.51
Percent Purity ≥97%
Chemical Name or Material 2,3,4-Tri-O-benzyl-beta-D-arabinopyranose
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.