Learn More
2,3,4-Trifluorobenzyl bromide, 97%, Thermo Scientific™
59.07 € - 242.00 €
Chemical Identifiers
CAS | 157911-55-2 |
---|---|
Molecular Formula | C7H4BrF3 |
Molecular Weight (g/mol) | 225.01 |
InChI Key | DGSXDQVPGXFOAN-UHFFFAOYSA-N |
Synonym | 2,3,4-trifluorobenzyl bromide, 1-bromomethyl-2,3,4-trifluorobenzene, 2,3,4-trifluorobenzylbromide, 6-bromomethyl-1,2,3-trifluorobenzene, benzene, 1-bromomethyl-2,3,4-trifluoro, alpha-bromo-2,3,4-trifluorotoluene, 2,3,4-trifluoro benzyl bromide, 2,3,4-trifluorophenyl methyl bromide, 4-bromomethyl-1,2,3-trifluorobenzene, 2, 3, 4-trifluorobenzyl bromideShow More |
PubChem CID | 2777050 |
IUPAC Name | 1-(bromomethyl)-2,3,4-trifluorobenzene |
SMILES | C1=CC(=C(C(=C1CBr)F)F)F |
Description
Chemical Identifiers
157911-55-2 | |
225.01 | |
2,3,4-trifluorobenzyl bromide, 1-bromomethyl-2,3,4-trifluorobenzene, 2,3,4-trifluorobenzylbromide, 6-bromomethyl-1,2,3-trifluorobenzene, benzene, 1-bromomethyl-2,3,4-trifluoro, alpha-bromo-2,3,4-trifluorotoluene, 2,3,4-trifluoro benzyl bromide, 2,3,4-trifluorophenyl methyl bromide, 4-bromomethyl-1,2,3-trifluorobenzene, 2, 3, 4-trifluorobenzyl bromideShow More | |
1-(bromomethyl)-2,3,4-trifluorobenzene |
C7H4BrF3 | |
DGSXDQVPGXFOAN-UHFFFAOYSA-N | |
2777050 | |
C1=CC(=C(C(=C1CBr)F)F)F |
Specifications
157911-55-2 | |
100.0 | |
90°C | |
96.5% min. (GC) | |
C7H4BrF3 | |
DGSXDQVPGXFOAN-UHFFFAOYSA-N | |
1-(bromomethyl)-2,3,4-trifluorobenzene | |
2777050 | |
97% | |
2, 3, 4-Trifluorobenzyl bromide |
96.0 | |
Colorless to Yellow | |
Authentic | |
Glass Bottle | |
2,3,4-trifluorobenzyl bromide, 1-bromomethyl-2,3,4-trifluorobenzene, 2,3,4-trifluorobenzylbromide, 6-bromomethyl-1,2,3-trifluorobenzene, benzene, 1-bromomethyl-2,3,4-trifluoro, alpha-bromo-2,3,4-trifluorotoluene, 2,3,4-trifluoro benzyl bromide, 2,3,4-trifluorophenyl methyl bromide, 4-bromomethyl-1,2,3-trifluorobenzene, 2, 3, 4-trifluorobenzyl bromide | |
C1=CC(=C(C(=C1CBr)F)F)F | |
225.01 | |
225.01 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- 2,3,4-Trifluorobenzyl bromide
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.