missing translation for 'onlineSavingsMsg'
Learn More
2-(3,5-Dimethyl-1H-pyrazol-1-yl)ethylamine, 97%, Thermo Scientific Chemicals Product Code.: 11409510

2-(3,5-Dimethyl-1H-pyrazol-1-yl)ethylamine, 97%, Thermo Scientific Chemicals

Product Code. 11409510
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11409510

Brand: Thermo Scientific Alfa Aesar H50333.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62821-88-9
Molecular Formula C7H13N3
Molecular Weight (g/mol) 139.20
MDL Number MFCD04967722
InChI Key FXLOXQBWUPCCDL-UHFFFAOYSA-N
Synonym 2-3,5-dimethyl-1h-pyrazol-1-yl ethanamine, 2-3,5-dimethyl-pyrazol-1-yl-ethylamine, 2-3,5-dimethyl-1h-pyrazol-1-yl ethan-1-amine, 2-3,5-dimethylpyrazol-1-yl ethanamine, 2-3,5-dimethylpyrazol-1-yl ethylamine, 2-3,5-dimethyl-1h-pyrazol-1-yl ethylamine, 1-2-aminoethyl-3,5-dimethylpyrazole, 2-3,5-dimethylpyrazolyl ethylamine, acmc-209v2h, 2-3,5-dimethyl-1-pyrazolyl ethanamineShow MoreShow Less
PubChem CID 1091687
SMILES CC1=NN(CCN)C(C)=C1

Specifications

Quantity 1 g
UN Number UN2735
Sensitivity Air and moisture sensitive
IUPAC Name 2-(3,5-dimethylpyrazol-1-yl)ethanamine
Formula Weight 139.2
Percent Purity 97%
Chemical Name or Material 2-(3,5-Dimethyl-1H-pyrazol-1-yl)ethylamine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP29H023 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50333.MD,H50333.03
Lot NumberB07S041 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50333.MD,H50333.03
Lot NumberJ04Q036 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50333.MD,H50333.03
Lot NumberA18S033 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50333.MD,H50333.03
Lot NumberE02S034 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50333.MD,H50333.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(3,5-Dimethyl-1H-pyrazol-1-yl)ethylamine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.