missing translation for 'onlineSavingsMsg'
Learn More
2-(3-Chlorophenyl)-1,3-thiazole-4-carboxylic acid, ≥97%, Thermo Scientific™ Product Code.: 10197282

2-(3-Chlorophenyl)-1,3-thiazole-4-carboxylic acid, ≥97%, Thermo Scientific™

Product Code. 10197282
1 g
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy

Product Code. 10197282

Brand: Thermo Scientific Maybridge MO01112DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 845885-82-7
Molecular Formula C10H6ClNO2S
Molecular Weight (g/mol) 239.67
MDL Number MFCD05664421
InChI Key DNSCDQVZFMLMAD-UHFFFAOYSA-N
Synonym 2-3-chlorophenyl-1,3-thiazole-4-carboxylic acid, 2-3-chlorophenyl thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-3-chlorophenyl, 2-3-chlorophenyl thiazole-4-carboxylicacid, 2-3-chlorophenyl-4-thiazolecarboxylic acid, 2-3-chlorophenyl-1 pound not3-thiazole-4-carboxylic acidShow MoreShow Less
PubChem CID 2794667
IUPAC Name 2-(3-chlorophenyl)-1,3-thiazole-4-carboxylic acid
SMILES OC(=O)C1=CSC(=N1)C1=CC(Cl)=CC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 239.68
Percent Purity 97%
Chemical Name or Material 2-(3-Chlorophenyl)-1, 3-thiazole-4-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(3-Chlorophenyl)-1,3-thiazole-4-carboxylic acid, ≥97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.