Learn More
2,3-Difluoro-6-methylpyridine, 98%, Thermo Scientific Chemicals Product Code.: 15481759
2,3-Difluoro-6-methylpyridine, 98%, Thermo Scientific Chemicals
Product Code. 15481759
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
Product Code. 15481759
Brand: Thermo Scientific Alfa Aesar H64514.MD
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1227579-04-5
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD16610828
InChI Key HGBWAUANMSZOIJ-UHFFFAOYSA-N
Synonym 2,3-difluoro-6-methyl-pyridine
PubChem CID 70680696
IUPAC Name 2,3-difluoro-6-methylpyridine
SMILES CC1=NC(=C(C=C1)F)F
Specifications
Color Colorless to Yellow
Quantity 250 mg
Formula Weight 129.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,3-Difluoro-6-methylpyridine
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2,3-Difluoro-6-methylpyridine, 98%, Thermo Scientific Chemicals >