missing translation for 'onlineSavingsMsg'
Learn More
2,4,6-Tri-tert-butylaniline, 95%, Thermo Scientific Chemicals Product Code.: 11304584

2,4,6-Tri-tert-butylaniline, 95%, Thermo Scientific Chemicals

Product Code. 11304584
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11304584

Brand: Thermo Scientific Alfa Aesar L01362.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,4,6-Tri-tert-butylnitrobenzene (bulky amine) is used in the synthesis of monomeric iminophosphorane

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Tri-tert-butylnitrobenzene (bulky amine) is used in the synthesis of monomeric iminophosphorane

Solubility
Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 961-38-6
Molecular Formula C18H31N
Molecular Weight (g/mol) 261.453
MDL Number MFCD00011645
InChI Key REJGDSCBQPJPQT-UHFFFAOYSA-N
Synonym 2,4,6-tri-tert-butylaniline, 2,4,6-tri-t-butylaniline, 2,4,6-tri tert-butyl aniline, aniline, tri-tert-butyl, 2,4,6-tri-tert-butylphenylamine, 2,4,6-tritert-butylphenylamine, benzenamine, 2,4,6-tris 1,1-dimethylethyl, aniline, 2,4,6-tri-tert-butyl, 2,4,6-tris tert-butyl phenylamine, pubchem16758Show MoreShow Less
PubChem CID 70402
IUPAC Name 2,4,6-tritert-butylaniline
SMILES CC(C)(C)C1=CC(=C(C(=C1)C(C)(C)C)N)C(C)(C)C

Specifications

Melting Point 144°C to 147°C
Quantity 1 g
UN Number UN2811
Beilstein 2272612
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 261.46
Percent Purity 95%
Chemical Name or Material 2,4,6-Tri-tert-butylaniline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242348 Certificate TypeCertificate of Analysis Date26/07/2022 Catalog NumberL01362.06,L01362.03
Lot Number10225971 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL01362.06,L01362.03
Lot NumberN22H063 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL01362.06,L01362.03
Lot NumberA08S033 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL01362.06,L01362.03
Lot Number10140611 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL01362.06,L01362.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4,6-Tri-tert-butylaniline, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.