missing translation for 'onlineSavingsMsg'
Learn More
2,4-Dimethyl-1,3-pentadiene, 98%, Thermo Scientific Chemicals Product Code.: 10616362

2,4-Dimethyl-1,3-pentadiene, 98%, Thermo Scientific Chemicals

Product Code. 10616362
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10616362

Brand: Thermo Scientific Acros 155360250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1000-86-8
Molecular Formula C7H12
Molecular Weight (g/mol) 96.17
MDL Number MFCD00008903
InChI Key CMSUNVGIWAFNBG-UHFFFAOYSA-N
Synonym 2,4-dimethyl-1,3-pentadiene, 1,1,3-trimethylbutadiene, 1,3-pentadiene, 2,4-dimethyl, acmc-20aoci, 1,3-trimethylbutadiene, 1,3-pentadiene,2,4-dimethyl, cmsunvgiwafnbg-uhfffaoysa, ch3 2c=chc ch3 =ch2, 1,3-pentadiene, 2,4-dimethyl-8ci 9ci
PubChem CID 66080
IUPAC Name 2,4-dimethylpenta-1,3-diene
SMILES CC(=CC(=C)C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7400g/mL
Boiling Point 94°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)2C=CHC(CH3)=CH2
Refractive Index 1.4400 to 1.442
Quantity 25 g
Beilstein 01, 257
Specific Gravity 0.74
Formula Weight 96.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Dimethyl-1, 3-pentadiene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462218 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog Number155360050,155360250,155361000
Lot NumberA0448544 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number155360050,155360250,155361000
Lot NumberA0417268 Certificate TypeCertificate of Analysis Date09/09/2022 Catalog Number155360050,155360250,155361000
Lot NumberA0403217 Certificate TypeCertificate of Analysis Date03/06/2021 Catalog Number155360050,155360250,155361000
compliance-icons compliance-icons
Product Identifier
  • 2,4-Dimethyl-1,3-pentadiene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4-Dimethyl-1,3-pentadiene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.