missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(trifluoromethyl)benzyl bromide, 98%, Thermo Scientific™

Product Code. 15404337
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15404337

Brand: Thermo Scientific Alfa Aesar H31711.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302911-98-4
Molecular Formula C9H5BrF6
Molecular Weight (g/mol) 307.033
MDL Number MFCD00799485
InChI Key CHSQGVCRNVEWIA-UHFFFAOYSA-N
Synonym 2,5-bis trifluoromethyl benzyl bromide, 2-bromomethyl-1,4-bis trifluoromethyl benzene, 2,5-bis trifluoromethyl benzylbromide, 2,5-bis trifluormethyl benzyl bromide, benzene,2-bromomethyl-1,4-bis trifluoromethyl, 2-2-bromoethyl-1,4-bis trifluoromethyl benzene, pubchem17545, acmc-1csxo, 1,4-bis trifluoromethyl-2-bromomethyl benzene
PubChem CID 2773199
IUPAC Name 2-(bromomethyl)-1,4-bis(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)CBr)C(F)(F)F

Specifications

Quantity 5 g
UN Number UN3265
Formula Weight 307.04
Percent Purity 98%
Chemical Name or Material 2,5-Bis(trifluoromethyl)benzyl bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.