missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

51.10 € - 77.89 €

Chemical Identifiers

CAS 89719-92-6
Molecular Formula C6H2Cl2F3N
Molecular Weight (g/mol) 215.98
MDL Number MFCD09749663
InChI Key GFTRYJQAASFSIB-UHFFFAOYSA-N
Synonym 2,5-dichloro-4-trifluoromethyl pyridine, pyridine, 2,5-dichloro-4-trifluoromethyl, acmc-20dwro
PubChem CID 26986016
IUPAC Name 2,5-dichloro-4-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC(Cl)=NC=C1Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15496247
SDS View Documents
Thermo Scientific Alfa Aesar
H33442.MD
250 mg N/A N/A N/A
15486247
SDS View Documents
Thermo Scientific Alfa Aesar
H33442.03
1 g
77.89 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-chloro-6-(trifluoromethyl)pyridine, 2-chloro-5-(trifluoromethyl)pyridine and 3-chloro-4-(trifluoromethyl)pyridine were each converted into the three possible carboxylic acids 2, 4, 6,8, 10, 12, 16, 17 and 20. This was achieved by employing several, but not all of the organometallic toolbox methods : transformation of a more basic organometallic species into a less basic isomer by transmetalation-equilibration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-chloro-6-(trifluoromethyl)pyridine, 2-chloro-5-(trifluoromethyl)pyridine and 3-chloro-4-(trifluoromethyl)pyridine were each converted into the three possible carboxylic acids 2, 4, 6,8, 10, 12, 16, 17 and 20. This was achieved by employing several, but not all of the organometallic “toolbox methods”: transformation of a more basic organometallic species into a less basic isomer by transmetalation-equilibration.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, aluminum. Protection under nitrogen and lower temperature is recommended(below 4 deg C).
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dichloro-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.