missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diphenyloxazole, Pure, Scintillation Counting, Fisher Chemical™

Product Code. 12698625 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
100g
5kg
This item is not returnable. View return policy

Product Code. 12698625

Brand: Fisher Chemical D/4865/65

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 92-71-7
Molecular Formula C15H11NO
Molecular Weight (g/mol) 221.26
MDL Number MFCD00005306
InChI Key CNRNYORZJGVOSY-UHFFFAOYSA-N
Synonym 2,5-diphenyloxazole, oxazole, 2,5-diphenyl, tritosol, 2,5-diphenyl-oxazole, ppo scintillator, dpo scintillator, usaf ek-6775, ppo van, ppo, ppo scintillator van
PubChem CID 7105
IUPAC Name 2,5-diphenyl-1,3-oxazole
SMILES O1C(=CN=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 70°C
Color White
Boiling Point 360°C
Packaging Plastic keg
Quantity 5 kg
Formula Weight 221.26g/mol
Physical Form Powder
Product Identifier
  • 2,5-Diphenyloxazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.