Learn More
2,6-Dichloro-3-nitrotoluene, 99%, Thermo Scientific Chemicals
Product Code. 11408887
Click to view available options
Quantity:
5 g
10 g
50 g
2,6-Dichloro-3-nitrotoluene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Dichloro-3-nitrotoluene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 29682-46-0
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.02
MDL Number MFCD00024185
InChI Key WBNZUUIFTPNYRN-UHFFFAOYSA-N
Synonym 2,6-dichloro-3-nitrotoluene, benzene, 1,3-dichloro-2-methyl-4-nitro, 2,4-dichloro-3-methylnitrobenzene, pubchem4540, acmc-209hao, ksc495g5p, wbnzuuiftpnyrn-uhfffaoysa, 2,4-dichloro-3-methyl-1-nitrobenzene, 1,3-dichloro-2-methyl-4-nitro-benzene
PubChem CID 97155
IUPAC Name 1,3-dichloro-2-methyl-4-nitrobenzene
SMILES CC1=C(Cl)C=CC(=C1Cl)[N+]([O-])=O
Specifications
Melting Point 53°C to 55°C
Quantity 50 g
UN Number UN2811
Beilstein 2259855
Solubility Information Insoluble in water.
Formula Weight 206.03
Percent Purity 99%
Chemical Name or Material 2,6-Dichloro-3-nitrotoluene