missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenylboronic acid, 97%, Thermo Scientific™

Product Code. 10517323
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10517323

Brand: Thermo Scientific Maybridge AC35911DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73852-17-2
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.81
MDL Number MFCD00064869
InChI Key CXDPUSMFYPQXCV-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzeneboronic acid, 2,6-dichlorophenyl boronic acid, 2,6-dichlorophenylboronicacid, 2,6-dichlorophenyl boranediol, boronic acid, 2,6-dichlorophenyl, pubchem7938, acmc-1bc3m, ksc489k8l, 2,6-dichloro-phenylboronic acid
PubChem CID 2734332
IUPAC Name (2,6-dichlorophenyl)boronic acid
SMILES OB(O)C1=C(Cl)C=CC=C1Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 190.82
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 2, 6-Dichlorophenylboronic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.