missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzyl chloride, 98%, Thermo Scientific Chemicals

Product Code. p-7024570
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11404060

Brand: Thermo Scientific Alfa Aesar A19472.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

2,6-Difluorobenzyl chloride is used as rufinamide intermediate. It is an important raw material and intermediate used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Difluorobenzyl chloride is used as rufinamide intermediate. It is an important raw material and intermediate used in organic synthesis.

Solubility
Soluble in acetone, DMSO and DMF. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 697-73-4
Molecular Formula C7H5ClF2
Molecular Weight (g/mol) 162.564
MDL Number MFCD00191346
InChI Key MJXRENZUAQXZGJ-UHFFFAOYSA-N
Synonym 2,6-difluorobenzyl chloride, 2-chloromethyl-1,3-difluorobenzene, 2,6-difluorobenzylchloride, benzene, 2-chloromethyl-1,3-difluoro, alpha-chloro-2,6-difluorotoluene, 2, 6-difluorobenzyl chloride, 1-chloromethyl-2,6-difluorobenzene, 2-chlormethyl-1,3-difluorbenzol, pubchem2178
PubChem CID 581570
IUPAC Name 2-(chloromethyl)-1,3-difluorobenzene
SMILES C1=CC(=C(C(=C1)F)CCl)F

Specifications

Melting Point 32°C to 35°C
Boiling Point 172°C to 173°C (640 mmHg)
Quantity 1 g
UN Number UN1760
Beilstein 243793
Solubility Information Soluble in acetone,DMSO and DMF. Insoluble in water.
Formula Weight 162.57
Percent Purity 98%
Chemical Name or Material 2,6-Difluorobenzyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.