missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Dihydroxy-4-methylpyridine, 98+%

Product Code. 15453797
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15453797

Brand: Alfa Aesar™ H27860.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
In order to synthesize macrocycles containing pyridine, 2,6-his-(bromomethyl)pyridine was prepared by bromonating 2,6-di (hydroxymethyl)pyridine.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4664-16-8
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD00040464
InChI Key JJHVYGVVMBYCMQ-UHFFFAOYSA-N
Synonym 6-hydroxy-4-methylpyridin-2 1h-one, 6-hydroxy-4-methyl-2-pyridone, 2,6-dihydroxy-4-methylpyridine, unii-0g0nnm04dt, 4-methylpyridine-2,6-diol, 0g0nnm04dt, 6-hydroxy-4-methyl-1,2-dihydropyridin-2-one, 2 1h-pyridinone, 6-hydroxy-4-methyl, acmc-209uzm, 4-methyl-2,6-dihydroxypyridine
PubChem CID 78395
IUPAC Name 6-hydroxy-4-methyl-1H-pyridin-2-one
SMILES CC1=CC(=O)NC(=C1)O

Specifications

Melting Point 195°C to 198°C
Quantity 250 mg
Solubility Information Soluble in water.
Formula Weight 125.13
Percent Purity ≥98%
Chemical Name or Material 2,6-Dihydroxy-4-methylpyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.