missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-2-heptanol, 99%, Thermo Scientific™

Product Code. 11471717
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13254-34-7
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.258
MDL Number MFCD00072198
InChI Key HGDVHRITTGWMJK-UHFFFAOYSA-N
Synonym 2,6-dimethyl-2-heptanol, 2-heptanol, 2,6-dimethyl, dimetol, freesiol, lolitol, dimethyl heptanol, acmc-20ao86, dimetol dimethyl heptanol, dsstox_cid_21424, dsstox_rid_79728
PubChem CID 83268
IUPAC Name 2,6-dimethylheptan-2-ol
SMILES CC(C)CCCC(C)(C)O

Specifications

Melting Point -10°C
Density 0.81
Boiling Point 179°C to 181°C
Flash Point 63°C (145°F)
Odor Characteristic
Refractive Index 1.426
Quantity 250 g
Formula Weight 144.25
Percent Purity 99%
Chemical Name or Material 2,6-Dimethyl-2-heptanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'