missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-3-cyano-4,5-di(fur-2-yl)furan, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Specifications
Specifications
| CAS Min % | 97.0 |
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 1 g |
| IUPAC Name | 2-amino-4,5-bis(furan-2-yl)furan-3-carbonitrile |
| Formula Weight | 240.22 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Amino-3-cyano-4, 5-di(fur-2-yl)furan |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction