missing translation for 'onlineSavingsMsg'
Learn More
2-Amino-3-cyano-4,5-di(fur-2-yl)furan, 97%, Thermo Scientific™ Product Code.: 10332255

2-Amino-3-cyano-4,5-di(fur-2-yl)furan, 97%, Thermo Scientific™

Product Code. 10332255
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 10332255

Brand: Thermo Scientific Maybridge NRB00495DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 24386-17-2
Molecular Formula C13H8N2O3
Molecular Weight (g/mol) 240.22
MDL Number MFCD00099027
InChI Key RDQSOEAGWZLHBE-UHFFFAOYSA-N
Synonym 2-amino-3-cyano-4,5-di fur-2-yl furan, 5'-amino-2,2':3',2-terfuran-4'-carbonitrile, 2-amino-3-cyano-4,5-di furyl furan, 2,2':3',2-terfuran-4'-carbonitrile,5'-amino, 2-amino-4,5-bis furan-2-yl furan-3-carbonitrile, 5'-amino-2'-furan-2-yl-2,3'-bifuran-4'-carbonitrile, 2-amino-4,5-di 2-furyl furan-3-carbonitrile, 5-amino-3-furan-2-yl-2,2'-bifuran-4-carbonitrile, aminocyanodifurylfuran
PubChem CID 714249
SMILES NC1=C(C#N)C(C2=CC=CO2)=C(O1)C1=CC=CO1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
IUPAC Name 2-amino-4,5-bis(furan-2-yl)furan-3-carbonitrile
Formula Weight 240.22
Percent Purity 97%
Chemical Name or Material 2-Amino-3-cyano-4, 5-di(fur-2-yl)furan
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Amino-3-cyano-4,5-di(fur-2-yl)furan, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.