missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-phenyl-3-furonitrile, 95%, Thermo Scientific™

Product Code. 10183444
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
This item is not returnable. View return policy

Product Code. 10183444

Brand: Thermo Scientific Maybridge AW01254EB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 14742-32-6
Molecular Formula C11H8N2O
Molecular Weight (g/mol) 184.20
MDL Number MFCD00463472
InChI Key UWTDTJWGYWUILP-UHFFFAOYSA-N
Synonym 2-amino-5-phenyl-3-furonitrile, 2-amino-3-cyano-5-phenylfuran, 2-amino-5-phenyl-3-furancarbonitrile, 3-furancarbonitrile, 2-amino-5-phenyl, maybridge1_000452, 2-amino-5-phenyl-furan-3-carbonitrile, 3-furancarbonitrile,2-amino-5-phenyl, 2-azanyl-5-phenyl-furan-3-carbonitrile
PubChem CID 693961
IUPAC Name 2-amino-5-phenylfuran-3-carbonitrile
SMILES NC1=C(C=C(O1)C1=CC=CC=C1)C#N

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 184.2
Percent Purity 95%
Chemical Name or Material 2-Amino-5-phenyl-3-furonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.