missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 15400646
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Reactant in the synthesis of isoxazoline indolizine amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of isoxazoline indolizine amides.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1171331-39-7
Molecular Formula C4H8ClF3N2O
Molecular Weight (g/mol) 192.566
MDL Number MFCD09944516
InChI Key DBNFKWRZLGVLSH-UHFFFAOYSA-N
Synonym 2-amino-n-2,2,2-trifluoroethyl acetamide hydrochloride, 2-amino-n-2,2,2-trifluoroethyl acetamide;hydrochloride, c4h7f3n2o.hcl, 2-amino-n-2,2,2-trifluoroethyl acetamide hcl
PubChem CID 42913698
IUPAC Name 2-amino-N-(2,2,2-trifluoroethyl)acetamide;hydrochloride
SMILES C(C(=O)NCC(F)(F)F)N.Cl

Specifications

Linear Formula H2NCH2(CO)NHCH2CF3·HCl
Quantity 5 g
Formula Weight 192.57
Percent Purity 97%
Chemical Name or Material 2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'