missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzenesulfonamide, 98%

Product Code. p-7026105
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11484987

Brand: Thermo Scientific Alfa Aesar B21912.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3306-62-5 | C6H8N2O2S | 172.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3306-62-5
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00007932
InChI Key YAZSBRQTAHVVGE-UHFFFAOYSA-N
Synonym orthanilamide, o-aminobenzenesulfonamide, o-sulfanilamide, benzenesulfonamide, 2-amino, 2-amino-benzenesulfonamide, benzenesulfonamide, o-amino, 2-aminobenzene-1-sulfonamide, o-aminobenzenesulphonamide, chembl6705, benzenesulfonamide, 2-amino-9ci
PubChem CID 72894
IUPAC Name 2-aminobenzenesulfonamide
SMILES C1=CC=C(C(=C1)N)S(=O)(=O)N

Specifications

Melting Point 153°C to 157°C
Odor Odorless
Quantity 5 g
Formula Weight 172.21
Percent Purity 98%
Chemical Name or Material 2-Aminobenzenesulfonamide
compliance-icons
Product Identifier
  • 2-Aminobenzenesulfonamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : o-Aminobenzenesulphonamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.