missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoxazolinone, 98%, Thermo Scientific Chemicals

Product Code. 11413977
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-49-4
Molecular Formula C7H5NO2
Molecular Weight (g/mol) 135.12
MDL Number MFCD00005716
InChI Key ASSKVPFEZFQQNQ-UHFFFAOYSA-N
Synonym 2-benzoxazolinone, benzoxazolinone, benzoxazolone, 2-hydroxybenzoxazole, 2-benzoxazolol, 2 3h-benzoxazolone, 1,3-benzoxazol-2 3h-one, 2-benzoxazolone, benzo d oxazol-2 3h-one, benzoxazolin-2-one
PubChem CID 6043
IUPAC Name 2,3-dihydro-1,3-benzoxazol-2-one
SMILES O=C1NC2=CC=CC=C2O1

Specifications

Melting Point 139°C to 140°C
Flash Point 160°C (320°F)
Quantity 25 g
Beilstein 119481
Formula Weight 135.12
Percent Purity 98%
Chemical Name or Material 2-Benzoxazolinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'