missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Bromo-5'-fluoroacetophenone, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2′-Bromo-5′-fluoroacetophenone is used in the synthesis of oxabicyclic compounds. It is also used in the enantioselective synthesis of chiral 3-aryl-1-indanone molecules.
Solubility
Soluble in methanol.
Notes
Light Sensitive. Store in cool, well ventilated area. Keep container tightly closed.
2′-Bromo-5′-fluoroacetophenone is used in the synthesis of oxabicyclic compounds. It is also used in the enantioselective synthesis of chiral 3-aryl-1-indanone molecules.
Solubility
Soluble in methanol.
Notes
Light Sensitive. Store in cool, well ventilated area. Keep container tightly closed.

Specifications
Specifications
Boiling Point | 144°C (18 mmHg) |
Refractive Index | 1.5525 |
Quantity | 5 g |
Synonym | 2'-bromo-5'-fluoroacetophenone, 1-2-bromo-5-fluorophenyl ethanone, 2-bromo-5-fluoroacetophenone, 1-2-bromo-5-fluorophenyl ethan-1-one, ethanone, 1-2-bromo-5-fluorophenyl, 1-2-bromo-5-fluoro-phenyl ethanone, ethanone,1-2-bromo-5-fluorophenyl, pubchem3214, acmc-1btyq, ksc499c4f |
Solubility Information | Soluble in methanol. |
Formula Weight | 217.04 |
Percent Purity | 98% |
Chemical Name or Material | 2'-Bromo-5'-fluoroacetophenone |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2'-Bromo-5'-fluoroacetophenone, 98%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction