missing translation for 'onlineSavingsMsg'
Learn More
2-Bromopyrimidine, 98+%, Thermo Scientific Chemicals Product Code.: 11454864

2-Bromopyrimidine, 98+%, Thermo Scientific Chemicals

Product Code. 11454864
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11454864

Brand: Thermo Scientific Alfa Aesar A13791.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Bromopyrimidine is used in the preparation of 2,7-bis(2-pyrimidyl)-9,9-dihexylfluorene by cross-coupling with 9,9-dihexylfluorene-2,7-diboronic acid. It undergoes microwave-assisted aminocarbonylation at C-2 with palladium and molybdenum hexacarbonyl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopyrimidine is used in the preparation of 2,7-bis(2-pyrimidyl)-9,9-dihexylfluorene by cross-coupling with 9,9-dihexylfluorene-2,7-diboronic acid. It undergoes microwave-assisted aminocarbonylation at C-2 with palladium and molybdenum hexacarbonyl.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids, acid chlorides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4595-60-2
Molecular Formula C4H3BrN2
Molecular Weight (g/mol) 158.99
MDL Number MFCD00014601
InChI Key PGFIHORVILKHIA-UHFFFAOYSA-N
Synonym pyrimidine, 2-bromo, 2-brompyrimidin, bromopyrimidine, 2-bromo pyrimidine, 2-bromo-pyrimidine, 2-bromanylpyrimidine, pubchem6901, acmc-1aqvo, 2-bromo-1,3-diazine, 2-bromopyrimidine
PubChem CID 78345
IUPAC Name 2-bromopyrimidine
SMILES BrC1=NC=CC=N1

Specifications

Melting Point 53°C to 57°C
Boiling Point 62°C to 64°C (1.5 mmHg)
Quantity 5 g
Beilstein 605945
Solubility Information Miscible with water.
Formula Weight 158.99
Percent Purity ≥98%
Chemical Name or Material 2-Bromopyrimidine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM23J037 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog NumberA13791.06,A13791.14,A13791.22
Lot NumberM23J036 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog NumberA13791.06,A13791.14,A13791.22
Lot NumberU30H017 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA13791.06,A13791.14,A13791.22
Lot Number10152765 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13791.06,A13791.14,A13791.22
Lot Number61000625 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13791.06,A13791.14,A13791.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromopyrimidine, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.