Learn More
2-Bromopyrimidine, 98+%, Thermo Scientific Chemicals
Description
2-Bromopyrimidine is used in the preparation of 2,7-bis(2-pyrimidyl)-9,9-dihexylfluorene by cross-coupling with 9,9-dihexylfluorene-2,7-diboronic acid. It undergoes microwave-assisted aminocarbonylation at C-2 with palladium and molybdenum hexacarbonyl.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Bromopyrimidine is used in the preparation of 2,7-bis(2-pyrimidyl)-9,9-dihexylfluorene by cross-coupling with 9,9-dihexylfluorene-2,7-diboronic acid. It undergoes microwave-assisted aminocarbonylation at C-2 with palladium and molybdenum hexacarbonyl.
Solubility
Miscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids, acid chlorides.

Specifications
Specifications
Melting Point | 53°C to 57°C |
Boiling Point | 62°C to 64°C (1.5 mmHg) |
Quantity | 25 g |
Beilstein | 605945 |
Solubility Information | Miscible with water. |
Formula Weight | 158.99 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Bromopyrimidine |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M23J037 | Certificate of Analysis | 27/01/2023 | A13791.06,A13791.14,A13791.22 |
M23J036 | Certificate of Analysis | 27/01/2023 | A13791.06,A13791.14,A13791.22 |
U30H017 | Certificate of Analysis | 12/04/2022 | A13791.06,A13791.14,A13791.22 |
10152765 | Certificate of Analysis | 22/11/2021 | A13791.06,A13791.14,A13791.22 |
61000625 | Certificate of Analysis | 22/11/2021 | A13791.06,A13791.14,A13791.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.